Assay Details
| Gene Symbol: Ns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169742.3 | NP_732199.2 | 4 - 5 | 1640 | 108 | |
| GenBank mRNA | BT050580.1 | - | 4 - 5 | 1752 | 108 | |
More Information
Set Membership: |
|

Ns1
Genomic Map