Assay Details
| Gene Symbol: Brf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170165.2 | NP_001163636.1 | 4 - 5 | 872 | 90 | |
| NM_142359.3 | NP_650616.2 | 4 - 5 | 872 | 90 | ||
| GenBank mRNA | AY051811.1 | - | 4 - 5 | 863 | 90 | |
More Information
Set Membership: |
|

Brf
Genomic Map