Assay Details
| Gene Symbol: cdm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275767.1 | NP_001262696.1 | 2 - 3 | 781 | 68 | |
| NM_142425.3 | NP_650682.1 | 2 - 3 | 781 | 68 | ||
| GenBank mRNA | AY061428.1 | - | 2 - 3 | 767 | 68 | |
More Information
Set Membership: |
|

cdm
Genomic Map