Assay Details
| Gene Symbol: MED17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142429.3 | NP_650686.1 | 1 - 2 | 147 | 57 | |
| GenBank mRNA | AF244916.1 | - | 1 - 2 | 115 | 57 | |
| AF289995.1 | - | 1 - 2 | 115 | 57 | ||
| AY118662.1 | - | 1 - 2 | 115 | 57 | ||
More Information
Set Membership: |
|

MED17
Genomic Map