Assay Details
| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142439.3 | NP_650696.1 | 2 - 3 | 386 | 59 | |
| GenBank mRNA | AY119152.1 | - | 2 - 3 | 383 | 59 | |
| BT029274.1 | - | 377 | 59 | |||
More Information
Set Membership: |
|

Mdh2
Genomic Map