Assay Details
| Gene Symbol: WRNexo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170175.2 | NP_001163646.1 | 4 - 5 | 510 | 75 | |
| NM_001300460.1 | NP_001287389.1 | 4 - 5 | 507 | 75 | ||
| NM_142458.4 | NP_650715.3 | 4 - 5 | 507 | 75 | ||
| GenBank mRNA | BT125935.1 | - | 4 - 5 | 483 | 75 | |
| EF680279.1 | - | 4 - 5 | 443 | 75 | ||
| EF680280.1 | - | 4 - 5 | 440 | 75 | ||
More Information
Set Membership: |
|

WRNexo
Genomic Map