Assay Details
| Gene Symbol: GluClalpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170185.3 | NP_001163656.2 | 4 - 5 | 896 | 100 | |
| NM_142570.4 | NP_650827.3 | 4 - 5 | 896 | 100 | ||
More Information
Set Membership: |
|

GluClalpha
Genomic Map