Assay Details
| Gene Symbol: GluClalpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014641.4 | NP_001014641.2 | 8 - 9 | 1498 | 73 | |
| NM_001038972.4 | NP_001034061.2 | 8 - 9 | 1495 | 73 | ||
| NM_001170183.2 | NP_001163654.1 | 7 - 8 | 1427 | 73 | ||
| NM_001170184.2 | NP_001163655.1 | 8 - 9 | 1495 | 73 | ||
| NM_001170185.3 | NP_001163656.2 | 8 - 9 | 1495 | 73 | ||
| NM_001275811.2 | NP_001262740.1 | 8 - 9 | 1498 | 73 | ||
| NM_001300479.1 | NP_001287408.1 | 8 - 9 | 1495 | 73 | ||
| NM_001300480.1 | NP_001287409.1 | 8 - 9 | 1495 | 73 | ||
| NM_001300481.1 | NP_001287410.1 | 8 - 9 | 1495 | 73 | ||
| NM_142570.4 | NP_650827.3 | 8 - 9 | 1495 | 73 | ||
| NM_169873.4 | NP_732447.2 | 8 - 9 | 1495 | 73 | ||
| GenBank mRNA | BT028797.1 | - | 5 - 6 | 697 | 73 | |
More Information
Set Membership: |
|

GluClalpha
Genomic Map