Assay Details
| Gene Symbol: Arc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142583.3 | NP_650840.1 | 2 - 3 | 389 | 64 | |
| GenBank mRNA | AY119269.1 | - | 2 - 3 | 378 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arc42| Gene Symbol: Arc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142583.3 | NP_650840.1 | 2 - 3 | 389 | 64 | |
| GenBank mRNA | AY119269.1 | - | 2 - 3 | 378 | 64 | |
Set Membership: |
|