Assay Details
| Gene Symbol: psidin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142601.2 | NP_650858.1 | 6 - 7 | 3010 | 75 | |
| GenBank mRNA | BT150356.1 | - | 6 - 7 | 3011 | 75 | |
More Information
Set Membership: |
|

psidin
Genomic Map