Assay Details
| Gene Symbol: Sirt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300493.1 | NP_001287422.2 | 3 - 4 | 399 | 74 | |
| NM_142623.4 | NP_650880.3 | 3 - 4 | 399 | 74 | ||
| GenBank mRNA | BT132954.1 | - | 3 - 4 | 394 | 74 | |
More Information
Set Membership: |
|

Sirt2
Genomic Map