Assay Details
| Gene Symbol: Pus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170193.1 | NP_001163664.1 | 3 - 4 | 879 | 64 | |
| NM_142642.2 | NP_650899.1 | 3 - 4 | 875 | 64 | ||
More Information
Set Membership: |
|

Pus1
Genomic Map