Assay Details
| Gene Symbol: AdSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142661.3 | NP_650918.1 | 2 - 3 | 410 | 67 | |
| GenBank mRNA | AY113428.1 | - | 2 - 3 | 445 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AdSS| Gene Symbol: AdSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142661.3 | NP_650918.1 | 2 - 3 | 410 | 67 | |
| GenBank mRNA | AY113428.1 | - | 2 - 3 | 445 | 67 | |
Set Membership: |
|