Assay Details
| Gene Symbol: Ice2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104382.2 | NP_001097852.1 | 3 | 2285 | 84 | |
| NM_142689.4 | NP_650946.3 | 1 | 2440 | 84 | ||
| GenBank mRNA | AY069681.1 | - | 1 | 2402 | 84 | |
| BT001660.1 | - | 3 | 2282 | 84 | ||
More Information
Set Membership: |
|

Ice2
Genomic Map