Assay Details
| Gene Symbol: Usp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260298.1 | NP_001247227.1 | 3 - 4 | 854 | 66 | |
| NM_142691.2 | NP_650948.2 | 7 - 8 | 2512 | 66 | ||
| GenBank mRNA | BT133006.1 | - | 3 - 4 | 854 | 66 | |
| BT133467.1 | - | 7 - 8 | 2512 | 66 | ||
More Information
Set Membership: |
|

Usp8
Genomic Map