Assay Details
| Gene Symbol: Nelf-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275875.1 | NP_001262804.1 | 5 - 6 | 3637 | 56 | |
| NM_001300509.1 | NP_001287438.1 | 5 - 6 | 3634 | 56 | ||
| NM_142701.2 | NP_650958.2 | 5 - 6 | 3637 | 56 | ||
| GenBank mRNA | BT150479.1 | - | 5 - 6 | 3654 | 56 | |
More Information
Set Membership: |
|

Nelf-A
Genomic Map