Assay Details
| Gene Symbol: CG15497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142719.2 | NP_650976.1 | 2 - 3 | 498 | 69 | |
| Gene Symbol: Fer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT029276.1 | - | 1562 | 69 | ||
More Information
Set Membership: |
|

CG15497
Genomic Map