Assay Details
| Gene Symbol: Burs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142726.2 | NP_650983.1 | 1 - 2 | 154 | 67 | |
| GenBank mRNA | AJ862523.1 | - | 1 - 2 | 121 | 67 | |
| AY672905.1 | - | 1 - 2 | 121 | 67 | ||
| BT122042.1 | - | 1 - 2 | 121 | 67 | ||
| BT132842.1 | - | 1 - 2 | 121 | 67 | ||
More Information
Set Membership: |
|

Burs
Genomic Map