Assay Details
| Gene Symbol: lsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142730.3 | NP_650987.1 | 2 - 3 | 726 | 63 | |
| GenBank mRNA | BT032686.1 | - | 2 - 3 | 640 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lsn| Gene Symbol: lsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142730.3 | NP_650987.1 | 2 - 3 | 726 | 63 | |
| GenBank mRNA | BT032686.1 | - | 2 - 3 | 640 | 63 | |
Set Membership: |
|