Assay Details
| Gene Symbol: dnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142738.3 | NP_650995.1 | 2 - 3 | 351 | 63 | |
| GenBank mRNA | AY070906.1 | - | 2 - 3 | 347 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dnd| Gene Symbol: dnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142738.3 | NP_650995.1 | 2 - 3 | 351 | 63 | |
| GenBank mRNA | AY070906.1 | - | 2 - 3 | 347 | 63 | |
Set Membership: |
|