Assay Details
| Gene Symbol: CG6015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142748.5 | NP_651005.1 | 1 - 2 | 924 | 59 | |
| GenBank mRNA | AY069490.1 | - | 1 - 2 | 914 | 59 | |
| Gene Symbol: CG45099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300529.1 | NP_001287458.1 | 1 - 2 | 924 | 59 | |
More Information
Set Membership: |
|

CG6015
Genomic Map