Assay Details
| Gene Symbol: Fadd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142749.3 | NP_651006.1 | 1 | 886 | 130 | |
| GenBank mRNA | AF222005.1 | - | 1 | 859 | 130 | |
| AF295103.1 | - | 1 | 1381 | 130 | ||
More Information
Set Membership: |
|

Fadd
Genomic Map