Assay Details
| Gene Symbol: CG6937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142809.4 | NP_651066.2 | 2 - 3 | 533 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG6937| Gene Symbol: CG6937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142809.4 | NP_651066.2 | 2 - 3 | 533 | 90 | |
Set Membership: |
|