Assay Details
| Gene Symbol: Gbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300548.1 | NP_001287477.1 | 1 | 569 | 75 | |
| NM_142848.3 | NP_651105.1 | 1 | 569 | 75 | ||
| GenBank mRNA | AY113571.1 | - | 1 | 567 | 75 | |
More Information
Set Membership: |
|

Gbp3
Genomic Map