Assay Details
Gene Symbol: Irk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142892.2 | NP_651149.2 | 4 - 5 | 1599 | 79 | |
NM_170076.2 | NP_732871.1 | 3 - 4 | 1305 | 79 | ||
GenBank mRNA | AY051445.1 | - | 4 - 5 | 1599 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
