Assay Details
| Gene Symbol: SPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142911.4 | NP_651168.1 | 3 - 4 | 812 | 59 | |
| GenBank mRNA | AY051638.1 | - | 3 - 4 | 797 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPE| Gene Symbol: SPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142911.4 | NP_651168.1 | 3 - 4 | 812 | 59 | |
| GenBank mRNA | AY051638.1 | - | 3 - 4 | 797 | 59 | |
Set Membership: |
|