Assay Details
| Gene Symbol: Hrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275953.1 | NP_001262882.1 | 2 - 3 | 1092 | 64 | |
| NM_142922.3 | NP_651179.1 | 1 - 2 | 828 | 64 | ||
| GenBank mRNA | AF181652.2 | - | 1 - 2 | 711 | 64 | |
More Information
Set Membership: |
|

Hrd3
Genomic Map