Assay Details
| Gene Symbol: Hrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275953.1 | NP_001262882.1 | 4 - 5 | 2006 | 83 | |
| NM_142922.3 | NP_651179.1 | 3 - 4 | 1742 | 83 | ||
| GenBank mRNA | AF181652.2 | - | 3 - 4 | 1625 | 83 | |
More Information
Set Membership: |
|

Hrd3
Genomic Map