Assay Details
| Gene Symbol: RpS19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275962.1 | NP_001262891.1 | 2 - 3 | 362 | 73 | |
| NM_142938.2 | NP_651195.1 | 2 - 3 | 369 | 73 | ||
More Information
Set Membership: |
|

RpS19b
Genomic Map