Assay Details
| Gene Symbol: CG13604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104428.1 | NP_001097898.1 | 2 - 3 | 475 | 76 | |
| NM_001275965.1 | NP_001262894.1 | 2 - 3 | 475 | 76 | ||
| NM_142945.3 | NP_651202.1 | 2 - 3 | 475 | 76 | ||
| GenBank mRNA | AY051411.1 | - | 2 - 3 | 475 | 76 | |
More Information
Set Membership: |
|

CG13604
Genomic Map