Assay Details
| Gene Symbol: CHORD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142969.4 | NP_651226.1 | 2 - 3 | 734 | 67 | |
| GenBank mRNA | AY070947.1 | - | 2 - 3 | 728 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHORD| Gene Symbol: CHORD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142969.4 | NP_651226.1 | 2 - 3 | 734 | 67 | |
| GenBank mRNA | AY070947.1 | - | 2 - 3 | 728 | 67 | |
Set Membership: |
|