Assay Details
| Gene Symbol: jnj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142971.3 | NP_651228.1 | 11 - 12 | 3123 | 93 | |
| GenBank mRNA | BT023747.1 | - | 11 - 12 | 3123 | 93 | |
More Information
Set Membership: |
|

jnj
Genomic Map