Assay Details
| Gene Symbol: jnj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142971.3 | NP_651228.1 | 2 - 3 | 376 | 67 | |
| GenBank mRNA | BT023747.1 | - | 2 - 3 | 376 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

jnj| Gene Symbol: jnj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142971.3 | NP_651228.1 | 2 - 3 | 376 | 67 | |
| GenBank mRNA | BT023747.1 | - | 2 - 3 | 376 | 67 | |
Set Membership: |
|