Assay Details
| Gene Symbol: CG6454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275986.1 | NP_001262915.1 | 6 - 7 | 2695 | 77 | |
| NM_142992.3 | NP_651249.2 | 6 - 7 | 4021 | 77 | ||
| NM_170141.3 | NP_732985.2 | 6 - 7 | 2695 | 77 | ||
More Information
Set Membership: |
|

CG6454
Genomic Map