Assay Details
| Gene Symbol: ana1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143011.3 | NP_651268.1 | 1 - 2 | 253 | 61 | |
| NM_170148.3 | NP_733008.2 | 1 - 2 | 337 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ana1| Gene Symbol: ana1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143011.3 | NP_651268.1 | 1 - 2 | 253 | 61 | |
| NM_170148.3 | NP_733008.2 | 1 - 2 | 337 | 61 | ||
Set Membership: |
|