Assay Details
| Gene Symbol: beta4GalT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143062.3 | NP_651319.2 | 2 - 3 | 836 | 72 | |
| GenBank mRNA | BT044519.1 | - | 2 - 3 | 819 | 72 | |
More Information
Set Membership: |
|

beta4GalT7
Genomic Map