Assay Details
| Gene Symbol: Mocs2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143083.3 | NP_651340.1 | 4 - 5 | 1274 | 64 | |
| GenBank mRNA | AY047520.1 | - | 4 - 5 | 1239 | 64 | |
| Gene Symbol: Mocs2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170249.1 | NP_001163720.1 | 4 - 5 | 1274 | 64 | |
More Information
Set Membership: |
|

Mocs2B
Genomic Map