Assay Details
| Gene Symbol: Sil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143099.4 | NP_651356.1 | 1 - 2 | 267 | 58 | |
| GenBank mRNA | AY058471.1 | - | 1 - 2 | 214 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sil1| Gene Symbol: Sil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143099.4 | NP_651356.1 | 1 - 2 | 267 | 58 | |
| GenBank mRNA | AY058471.1 | - | 1 - 2 | 214 | 58 | |
Set Membership: |
|