Assay Details
| Gene Symbol: Npl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170260.2 | NP_001163731.1 | 3 - 4 | 467 | 76 | |
| NM_143150.4 | NP_651407.3 | 3 - 4 | 661 | 76 | ||
| NM_170231.3 | NP_733110.2 | 3 - 4 | 467 | 76 | ||
| GenBank mRNA | AY058286.2 | - | 3 - 4 | 661 | 76 | |
More Information
Set Membership: |
|

Npl4
Genomic Map