Assay Details
| Gene Symbol: Vps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143198.2 | NP_651455.1 | 1 - 2 | 410 | 71 | |
| GenBank mRNA | BT003747.1 | - | 1 - 2 | 406 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps2| Gene Symbol: Vps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143198.2 | NP_651455.1 | 1 - 2 | 410 | 71 | |
| GenBank mRNA | BT003747.1 | - | 1 - 2 | 406 | 71 | |
Set Membership: |
|