Assay Details
| Gene Symbol: CG6154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143214.2 | NP_651471.2 | 6 - 7 | 1463 | 91 | |
| NM_170267.2 | NP_733146.2 | 6 - 7 | 1156 | 91 | ||
| GenBank mRNA | BT010258.1 | - | 6 - 7 | 1463 | 91 | |
| BT031191.1 | - | 6 - 7 | 1159 | 91 | ||
More Information
Set Membership: |
|

CG6154
Genomic Map