Assay Details
| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276069.1 | NP_001262998.1 | 2 - 3 | 236 | 83 | |
| NM_143252.4 | NP_651509.2 | 2 - 3 | 248 | 83 | ||
| GenBank mRNA | AJ133736.1 | - | 1 - 2 | 160 | 83 | |
| BT128781.1 | - | 2 - 3 | 243 | 83 | ||
More Information
Set Membership: |
|

SPARC
Genomic Map