Assay Details
| Gene Symbol: grass | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143286.3 | NP_651543.1 | 3 - 4 | 561 | 56 | |
| NM_170318.3 | NP_733197.1 | 2 - 3 | 635 | 56 | ||
| GenBank mRNA | BT003238.1 | - | 2 - 3 | 619 | 56 | |
More Information
Set Membership: |
|

grass
Genomic Map