Assay Details
| Gene Symbol: TTLL6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143292.2 | NP_651549.1 | 2 - 3 | 896 | 105 | |
| GenBank mRNA | AY089310.1 | - | 2 - 3 | 884 | 105 | |
More Information
Set Membership: |
|

TTLL6B
Genomic Map