Assay Details
| Gene Symbol: bigmax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143299.3 | NP_651556.2 | 1 - 2 | 198 | 103 | |
| GenBank mRNA | AY118474.1 | - | 1 - 2 | 167 | 103 | |
More Information
Set Membership: |
|

bigmax
Genomic Map