Assay Details
| Gene Symbol: CG3339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043297.3 | NP_001036762.2 | 26 - 27 | 11153 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG3339| Gene Symbol: CG3339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043297.3 | NP_001036762.2 | 26 - 27 | 11153 | 61 | |
Set Membership: |
|