Assay Details
| Gene Symbol: CG42534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260404.1 | NP_001247333.1 | 1 - 2 | 66 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG42534| Gene Symbol: CG42534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260404.1 | NP_001247333.1 | 1 - 2 | 66 | 93 | |
Set Membership: |
|