Assay Details
| Gene Symbol: CG42534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260404.1 | NP_001247333.1 | 2 - 3 | 338 | 80 | |
| NM_143303.3 | NP_651560.2 | 2 - 3 | 713 | 80 | ||
More Information
Set Membership: |
|

CG42534
Genomic Map