Assay Details
| Gene Symbol: wdb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382180.1 | NP_001368946.1 | 6 - 7 | 1928 | 77 | |
| NM_143312.4 | NP_651569.1 | 6 - 7 | 2088 | 77 | ||
| NM_170336.3 | NP_733215.1 | 6 - 7 | 2249 | 77 | ||
| NM_170337.3 | NP_733216.1 | 6 - 7 | 1928 | 77 | ||
| NM_170338.3 | NP_733217.1 | 7 - 8 | 2097 | 77 | ||
| NM_170339.3 | NP_733218.1 | 7 - 8 | 2257 | 77 | ||
| NM_170340.2 | NP_733219.1 | 6 - 7 | 2341 | 77 | ||
| GenBank mRNA | AF145696.1 | - | 6 - 7 | 2215 | 77 | |
More Information
Set Membership: |
|

wdb
Genomic Map