Assay Details
| Gene Symbol: ALiX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143325.4 | NP_651582.1 | 3 - 4 | 743 | 63 | |
| GenBank mRNA | AY069534.1 | - | 3 - 4 | 841 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALiX| Gene Symbol: ALiX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143325.4 | NP_651582.1 | 3 - 4 | 743 | 63 | |
| GenBank mRNA | AY069534.1 | - | 3 - 4 | 841 | 63 | |
Set Membership: |
|